Learn More
X5 75ML DIGESTION TUBE .
Designed for use with blockheaters
Marke: Quickfit TJ75/26/3
Weitere Details : Netto-Gewicht : 0.56000kg
Beschreibung
- Contains Quickfit precision-ground socket to accept air condensers
- Manufactured from borosilicate glass for highest chemical and thermal durability