missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridaben-d13, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Pyridaben D13 (tert-butylphenyl D13) |
| CAS | 2468638-05-1 |
| Hinweise zur Reinheitsqualität | HPLC |
| Summenformel | C19 D13 H12 Cl N2 O S |
| Menge | 10 mg |
| Synonym | Pyridaben D13 (tert-butylphenyl D13) |
| InChI-Formel | InChI=1S/C19H25ClN2OS/c1-18(2,3)14-9-7-13(8-10-14)12-24-15-11-21-22(19(4,5)6)17(23)16(15)20/h7-11H,12H2,1-6H3/i1D3,2D3,3D3,7D,8D,9D,10D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1CSC2=C(Cl)C(=O)N(N=C2)C(C)(C)C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC-Name | 2-tert-Butyl-4-chlor-5-[[2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]phenyl]methylsulfanyl]pyridazin-3-on |
| Molekulargewicht (g/mol) | 378.013 |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?