missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QRSL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15926-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Allows the formation of correctly charged Gln-tRNA(Gln) through the transamidation of misacylated Glu-tRNA(Gln) in the mitochondria. The reaction takes place in the presence of glutamine and ATP through an activated gamma-phospho-Glu- tRNA(Gln).Spezifikation
QRSL1 | |
Polyclonal | |
Unconjugated | |
QRSL1 | |
GatA, QRSL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309911, 55278, 76563 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5FWT5, Q9CZN8, Q9H0R6 | |
Qrsl1 | |
QRSL1 Fusion Protein Ag8756 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |