Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Marke: Quickfit EX13/25
Weitere Details : Netto-Gewicht : 0.01100kg
Beschreibung
- Designed to support Raschig rings within plain fractionating columns
- Support is passed through socket of column and rests at the constriction above the cone
- Manufactured from chemically resistant borosilicate glass