Learn More
Quickfit™ Borosilicate Glass Florentine Flask
Allows easy removal of solid residues
Marke: Quickfit FD250/3
Weitere Details : Netto-Gewicht : 0.14000kg
Beschreibung
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues