Learn More
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
Marke: Quickfit FF500/5S
Weitere Details : Netto-Gewicht : 1.11000kg
Beschreibung
- Complies with ISO 1773 and ISO 4797
- With highest quality Quickfit™ ground glass socket
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Flat base allows use with hotplates