Learn More
Quickfit™ Borosilicate Glass Single Neck Pear Shaped Flask
Allows distillation down to small quantities.
Marke: Quickfit FP5/1
Weitere Details : Netto-Gewicht : 0.01100kg
Beschreibung
- With highest quality Quickfit™ ground glass socket
- Ideal for semi-micro distillations
- Pear shaped design allows distillation down to small quantities with minimum hold-up, and reduced losses from thermal decomposition
- Borosilicate glass for chemical resistance and thermal performance