Learn More
BODY FOR DILATOMETERS .
Marke: Quickfit WE12/1
Weitere Details : Netto-Gewicht : 0.07000kg
Beschreibung
- For measuring the expansion of edible oils and fats over the temperature range within which melting takes place
- Dilatometer body is graduated with an arbitrary scale
- Interchangeable stopper has open top to allow weighting with steel balls etc