Learn More
Alfa Aesar™ (R)-(+)-1-(1-naphthyl)ethylamin, ChiPros 99+ %, ee 98+ %
Marke: Alfa Aesar™ L19300.06
Weitere Details : CAS : 3886-70-2 Netto-Gewicht : 0.00500kg Transport : UN number : 2922 Chem class : 8 Pack group : III
Menge | 5g |
---|
Chemische Identifikatoren
3886-70-2 | |
171.243 | |
RTCUCQWIICFPOD-SECBINFHSA-N | |
2724264 | |
CC(C1=CC=CC2=CC=CC=C21)N |
C12H13N | |
MFCD00064114 | |
r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine | |
(1R)-1-Naphthalen-1-ylethanamin |
Spezifikation
(R)-(+)-1-(1-Naphthyl)ethylamine | |
>110°C (230°F) | |
+55° (c=2 in Ethanol) | |
Air sensitive | |
5g | |
ee ≥98 % | |
MFCD00064114 | |
2208025 | |
RTCUCQWIICFPOD-SECBINFHSA-N | |
(1R)-1-Naphthalen-1-ylethanamin | |
2724264 | |
≥99% |
1.06 | |
Amine-like | |
1.623 | |
152°C to 153°C (11mmHg) | |
3886-70-2 | |
C12H13N | |
UN2922 | |
r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine | |
CC(C1=CC=CC2=CC=CC=C21)N | |
171.243 | |
171.24 |