missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure, Thermo Scientific Chemicals

Catalog Number p-7030487
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
103733-65-9
C10H11NO2
177.20
MFCD00144038
BWKMGYQJPOAASG-UHFFFAOYNA-N
r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
712398
OC(=O)C1CC2=CC=CC=C2CN1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
103733-65-9
C10H11NO2
177.20
MFCD00144038
BWKMGYQJPOAASG-UHFFFAOYNA-N
r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-d-tic-oh, d-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, r-+-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3-isoquinolinecarboxylic acid, 1,2,3,4-tetrahydro-, 3r, 3r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, 3r-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid, r-1,2,3,4-tetrahydro-3-isoquinolinecarboxylicacid
712398
OC(=O)C1CC2=CC=CC=C2CN1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (R)-(+)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Menge 250 mg
Optische Drehung +163° (c=2 in 1N NaOH)
IUPAC-Name 1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Formelmasse 177.2

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'