missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(di-3,5-Xylphosphino)-1,1'-Binaphthyl, 98 %, Thermo Scientific™

Catalog Number p-7030137
Click to view available options
Menge:
100 mg
500 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
137219-86-4
C52H48P2
734.904
MFCD01630821
MXGXXBYVDMVJAO-UHFFFAOYSA-N
s-dm-binap, dm-binap, r-dm-binap, s---2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, s-3,5-xylyl-binap, r-+-2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, 3,5-xylyl-binap, s-xylyl-binap, r-2,2'-bis bis 3,5-dimethylphenyl phosphino-1,1'-binaphthalene, r-+-xylbinap
4189905
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphan
CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
137219-86-4
C52H48P2
734.904
MFCD01630821
MXGXXBYVDMVJAO-UHFFFAOYSA-N
s-dm-binap, dm-binap, r-dm-binap, s---2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, s-3,5-xylyl-binap, r-+-2,2'-bis di 3,5-xylyl phosphino-1,1'-binaphthyl, 3,5-xylyl-binap, s-xylyl-binap, r-2,2'-bis bis 3,5-dimethylphenyl phosphino-1,1'-binaphthalene, r-+-xylbinap
4189905
[1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphan
CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material (R)-(+)-2,2'-Bis(di-3,5-xylylphosphino)-1,1'-binaphthyl
Schmelzpunkt 201°C to 203°C
Menge 100 mg
Formelmasse 734.91
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'