Learn More
(R)-2-Phenylpropanoic acid, 98%, BLD Pharm™
Product Code. 31449368
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31449368 100 g
31443828 25 g
31434632 10 g
31442337 1 g
31435191 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31449368 Supplier BLD Pharm Supplier No. BD11237100g
 Restricted Item
Verfügbar ab: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 7782-26-5
Molecular Formula C9H10O2
MDL Number MFCD00063140
Quantity 100 g
SMILES C[C@H](C1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 150.17
Percent Purity 98%
Chemical Name or Material (R)-2-Phenylpropanoic acid