missing translation for 'onlineSavingsMsg'
Learn More

R 568 hydrochloride, Tocris Bioscience™

Positive allosteric modulator of the human calcium-sensing receptor

Marke:  Tocris (Bio-Techne) 381550

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413644

  • 778.20 € / 50 Milligramm


Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Positive allosteric modulator and allosteric agonist of the human calcium-sensing receptor (hCaSR). Exhibits calcimimetic activity. Has the ability to restore function to hCaSR mutants that cause familial hypocalciuric hypercalcemia (FHH) and neonatal severe hyperparathyroidism (NSHPT). Exerts a suppressive effect on parathyroid (PT) hormone secretion; inhibits PT cell proliferation in rats with renal insufficiency.
Spezifikation

Chemische Identifikatoren

177172-49-5
340.288
r 568 hydrochloride, tecalcet hydrochloride, tecalcet hcl, norcalcin, unii-3hp28r98lc, tecalcet hydrochloride usan, r-3-2-chlorophenyl-n-1-3-methoxyphenyl ethyl propan-1-amine hydrochloride, tecalcet hydrochloride usan, tecalcet hydrochloride, 3-2-chlorophenyl propyl 1r-1-3-methoxyphenyl ethyl amine hydrochloride
3-(2-chlorophenyl)-N-[(1R)-1-(3-methoxyphenyl)ethyl]propan-1-amine;hydrochloride
C18H23Cl2NO
YJXUXANREVNZLH-PFEQFJNWSA-N
158796
CC(C1=CC(=CC=C1)OC)NCCCC2=CC=CC=C2Cl.Cl

Spezifikation

R 568 hydrochloride
177172-49-5
r 568 hydrochloride, tecalcet hydrochloride, tecalcet hcl, norcalcin, unii-3hp28r98lc, tecalcet hydrochloride usan, r-3-2-chlorophenyl-n-1-3-methoxyphenyl ethyl propan-1-amine hydrochloride, tecalcet hydrochloride usan, tecalcet hydrochloride, 3-2-chlorophenyl propyl 1r-1-3-methoxyphenyl ethyl amine hydrochloride
CC(C1=CC(=CC=C1)OC)NCCCC2=CC=CC=C2Cl.Cl
340.288
340.29
50mg
C18H23Cl2NO
YJXUXANREVNZLH-PFEQFJNWSA-N
3-(2-chlorophenyl)-N-[(1R)-1-(3-methoxyphenyl)ethyl]propan-1-amine;hydrochloride
158796
>99%
Videos