Learn More
R&D Systems™ Amphibian TGF-beta 5 Recombinant Protein
Marke:  R&D Systems™ 245-B5-002
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411254
  • 333.97 € / 2 Mikrogramm
Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to inhibit the IL-4-dependent proliferation of HT-2 mouse T cells. Tsang, M. et al. (1995) Cytokine 7:389. The ED 50 for this effect is 0.01-0.03 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
12.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Amphibian
Unlabeled
Reconstituted at 20 μg/mL in sterile 4 mM HCl containing at least 0.1% human or bovine serum albumin.
397778
TGF-beta 5
2μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.