Learn More
R&D Systems™ b-AP15/VLX1500
Specifically inhibits the 19S regulatory particle-associated deubiquitinases USP14 and UCHL5, resulting in the accumulation of intracellular polyubiquitin
N.V.
Beschreibung
Beschreibung
b-AP15 has been shown to inhibit tumor progression in at least four solid tumor models, and also inhibits organ infiltration in an acute myeloid leukemia (AML) model.
  • Potent, reversible, cell-permeable inhibitor
  • Purity >97% by HPLC
  • Decreases HCT-116 cell viability at 72 hours (EC50 = 0.4μM)