Learn More
R&D Systems™ b-AP15/VLX1500
Specifically inhibits the 19S regulatory particle-associated deubiquitinases USP14 and UCHL5, resulting in the accumulation of intracellular polyubiquitin
Marke:  R&D Systems™ I-197-01M
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13417425
  • 101.25 € / 1 Milligramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
b-AP15 has been shown to inhibit tumor progression in at least four solid tumor models, and also inhibits organ infiltration in an acute myeloid leukemia (AML) model.
  • Potent, reversible, cell-permeable inhibitor
  • Purity >97% by HPLC
  • Decreases HCT-116 cell viability at 72 hours (EC50 = 0.4μM)
Spezifikation
Spezifikation
19S proteasome inhibitor
419.39
Store the unopened product at 2 - 8° C. Do not use past expiration date.
19S proteasome
1mg
Videos