missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Feline GM-CSF Recombinant Protein

Marke:  R&D Systems™ 987-FL-010

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416494

  • 442.02 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ala18-Lys144 (Met36Ile, Thr56Ala & Lys126Asn), with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 493805
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
14.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Yes
493805
GM-CSF
10μg
Unlabeled
Videos