missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Feline IL-10 Recombinant Protein

Marke:  R&D Systems™ 736-FL-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416154

  • 304.93 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 26-03-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ser19-Ile178 & Ser26-Ile178
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4.
Entrez Gene ID: 493683
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in NaH2PO4.
18.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Feline
Unlabeled
493683
IL-10
10μg
Noncovalently-linked homodimer
Yes
Videos