missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human 4-1BB Ligand/TNFSF9 Recombinant Protein

Marke:  R&D Systems™ 2295-4L-025

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13410964

  • 366.97 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Arg71-Glu254, with an N-terminal Met and a 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, NaCl and PEG.
Entrez Gene ID: 8744
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS, NaCl and PEG with BSA as a carrier protein.
20.4 kDa
25μg
4-1BB Ligand/TNFSF9
Yes
8744
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Human
Videos