missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Active Caspase-2 Recombinant Protein

Marke:  R&D Systems™ 702-C2-010

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416034

  • 448.52 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Gly170-Asp333 & Ala348-Tyr452
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12 kDa & 18 kDa
Formulation: Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
Entrez Gene ID: 835
Regulatory Status: RUO

Spezifikation

Spezifikation

Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose with BSA as a carrier protein.
12 kDa & 18 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
835
Active Caspase-2
10μg
Unlabeled
Videos