missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Angiogenin Recombinant Protein

Marke:  R&D Systems™ 265-AN-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411674

  • 503.12 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ribonucleolytic activity toward yeast tRNA. Lee and Vallee (1989) Biochem. Biophys. Res. Commun. 161:121. Angiogenin produces a delta Abs 260µg >1.0 in two hours, as measured under the described conditions.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
14 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
283
Angiogenin
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos