missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Angiopoietin-4 Recombinant Protein

Marke:  R&D Systems™ 964-AN-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416464

  • 460.22 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived
Purity: 85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 57 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
Entrez Gene ID: 51378
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl.
57 kDa (monomer)
85%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
51378
Angiopoietin-4
25μg
Oligomer
Yes
Videos