missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-10 Recombinant Protein

Marke:  R&D Systems™ 2926-BP-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412064

  • 633.57 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to induce alkaline phosphatase production by MC3T3-E1 mouse preosteoblast cells. The ED 50 for this effect is 15-60 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
12.2 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked homodimer
Yes
27302
BMP-10
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Videos