missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human BMP-4/BMP-7 Heterodimer Recombinant Protein

Marke:  R&D Systems™ 3727-BP-010

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413504

  • 562.92 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ser293-Arg408
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.3 kDa (BMP-4), 15.8 kDa (BMP-7)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 652
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
13.3 kDa (BMP-4), 15.8 kDa (BMP-7)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
652
BMP-4/BMP-7 Heterodimer
10μg
Disulfide-linked heterodimer
Yes
Videos