missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Caspase-7 Recombinant Protein

Marke:  R&D Systems™ 823-C7-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416354

  • 372.25 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ala24-Asp198 (p20) & Ala207-Gln303 (p11)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 20 kDa (p20) & 11 kDa (p11)
Formulation: Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
Entrez Gene ID: 840
Regulatory Status: RUO

Spezifikation

Spezifikation

Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
20 kDa (p20) & 11 kDa (p11)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
840
Caspase-7
10μg
Unlabeled
Videos