missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL14a/HCC-1 aa 20-93 Recombinant Protein

Marke:  R&D Systems™ 324-HC-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412654

  • 171.60 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to chemoattract 2-day cultured human monocytes. The ED 50 for this effect is 5-15 µg/mL. Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR1. The ED 50 for this effect is 0.2-0.8 µg/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
8.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6358
CCL14a/HCC-1 aa 20-93
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos