missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL14a/HCC-1 aa 20-93 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Konjugat | Unconjugated |
---|---|
Gen-ID (Entrez) | 6358 |
Molekulargewicht | 8.7 kDa |
Name | CCL14a/HCC-1 aa 20-93 |
Rekombinant | Yes |
Beschreibung
The Recombinant Human CCL14a/HCC-1 Protein is derived from E. coli. The Recombinant Human CCL14a/HCC-1 Protein has been validated for the following applications: Bioactivity.Spezifikation
Unconjugated | |
8.7 kDa | |
Yes |
6358 | |
CCL14a/HCC-1 aa 20-93 | |
E. coli-derived human CCL14a/HCC-1 protein Thr20-Asn93 |