missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL2/JE/MCP-1 Recombinant Protein

Marke:  R&D Systems™ 279-MC-010/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13411864

  • 319.45 € / 10 Mikrogramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR2A. The ED 50 for this effect is 5-30 ng/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
8.7 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6347
CCL2/JE/MCP-1
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos