Learn More
R&D Systems™ Human CCL4/MIP-1 beta Recombinant Protein
Marke:  R&D Systems™ 271-BME-050
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411804
  • 462.82 € / 50 Mikrogramm
Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR5. The ED 50 for this effect is 1-6 ng/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.8 kDa & 7.3 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6351
CCL4/MIP-1 beta
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.