Learn More
R&D Systems™ Human CCL4/MIP-1 beta Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Konjugat Unconjugated
Gen-ID (Entrez) 6351
Name CCL4/MIP-1 beta
Rekombinant Yes
Quelle E. coli-derived human CCL4/MIP-1 beta protein Ala24-Asn92 (major); Pro30-Asn92 (Minor)
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Recombinant Human CCL4/MIP-1 beta Protein is derived from E. coli. The Recombinant Human CCL4/MIP-1 beta Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Unconjugated
CCL4/MIP-1 beta
E. coli-derived human CCL4/MIP-1 beta protein Ala24-Asn92 (major); Pro30-Asn92 (Minor)
6351