missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL7/MCP-3/MARC Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 6354
Molekulargewicht 9 kDa
Name CCL7/MCP-3/MARC
Rekombinant Yes
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Human CCL7/MCP-3 Protein is derived from E. coli. The Recombinant Human CCL7/MCP-3 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
9 kDa
Yes
6354
CCL7/MCP-3/MARC
E. coli-derived human CCL7/MCP-3/MARC protein Gln34-Leu109
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen