Learn More
R&D Systems™ Human CCL7/MCP-3/MARC Recombinant Protein
Marke:  R&D Systems™ 282-P3-010
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411914
  • 315.49 € / 10 Mikrogramm
Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to chemoattract 2-day cultured human monocytes. Matsushima, K. et al. (1989) J. Exp. Med. 169:1485. The ED 50 for this effect is 0.02-0.08 µg/mL. Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR2A. The ED 50 for this effect is 0.1-0.5 µg/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6354
CCL7/MCP-3/MARC
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.