missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL8/MCP-2 Recombinant Protein

Marke:  R&D Systems™ 281-CP-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Ver más versiones de este producto

Código de producto. 13411894

  • 275.89 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 27-06-2019
Añadir a la cesta

Descripción

Descripción

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CCR5. The ED 50 for this effect is 0.01-0.06 µg/mL. Measured by its ability to chemoattract THP-1 human acute monocytic leukemia cells. The ED 50 for this effect is 0.03-0.1 µg/mL.
Especificaciones

Especificaciones

Lyophilized from a 0.2μm filtered solution in PBS.
9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
6355
CCL8/MCP-2
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos