missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL8/MCP-2 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 6355
Molekulargewicht 9 kDa
Name CCL8/MCP-2
Rekombinant Yes
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Human CCL8/MCP-2 Protein is derived from E. coli. The Recombinant Human CCL8/MCP-2 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
9 kDa
Yes
6355
CCL8/MCP-2
E. coli-derived human CCL8/MCP-2 protein Gln24-Pro99
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen