missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CX3CL1/Fractalkine Full Length Recombinant Protein

Marke:  R&D Systems™ 365-FR-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413374

  • 481.02 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Gln25-Arg339, with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 6376
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
35 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
6376
CX3CL1/Fractalkine Full Length
25μg
Unlabeled
Videos