missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL13/BLC/BCA-1 Recombinant Protein

Marke:  R&D Systems™ 801-CX-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416334

  • 494.02 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CXCR5. The ED 50 for this effect is 0.005-0.02 µg/mL.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
8.7 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
10563
CXCL13/BLC/BCA-1
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos