missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL7/NAP-2 Recombinant Protein

Marke:  R&D Systems™ 393-NP-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413854

  • 418.62 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 22-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Ala59-Asp128
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 5473
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.6 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
5473
CXCL7/NAP-2
50μg
Unlabeled
Videos