missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Endoglin/CD105 Recombinant Protein

For use in cell or tissue culture applications.

Marke:  R&D Systems™ 1097-EN-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418483

  • 487.52 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Source: Mouse myeloma cell line, NS0-derived, Glu26-Gly586
  • Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular Weight: 61 kDa
  • Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
  • Entrez Gene ID: 2022
  • Regulatory Status: RUO
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
61 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Yes
2022
Endoglin/CD105
25μg
Unlabeled
Videos