Learn More
R&D Systems™ Human/Feline CXCL12/SDF-1 beta aa 19-93 Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 2716-SD-025
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411794
  • 279.30 € / 25 Mikrogramm
Voraussichtlicher Liefertermin 22-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
The Recombinant Human/Feline CXCL12/SDF-1 beta (aa 19-93) is derived from E. coli. The Recombinant Human/Feline CXCL12/SDF-1 beta (aa 19-93) has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human/Feline
Unlabeled
6387
CXCL12/SDF-1 beta aa 19-93
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes