Learn More
R&D Systems™ Human/Feline CXCL12/SDF-1 beta aa 19-93 Recombinant Protein
Marke:  R&D Systems™ 2716-SD-025
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411794
  • 250.81 € / 25 Mikrogramm
Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CXCR4. The ED 50 for this effect is 0.15-0.6 µg/mL.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
8.8 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human/Feline
Unlabeled
6387
CXCL12/SDF-1 beta aa 19-93
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.