Learn More
R&D Systems™ Human FGF-21 Recombinant Protein
Marke:  R&D Systems™ 2539-FG-025
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411434
  • 390.01 € / 25 Mikrogramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with human FGF RIIIc. The ED 50 for this effect is 0.06-0.4 µg/mL in the presence of Recombinant Mouse Klotho beta (Catalog # 2619-KB ).
Spezifikation
Spezifikation
Lyophilized from a 0.2 μm filtered solution in MES, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein.
20.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
26291
FGF-21
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.