missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Follistatin 288 Recombinant Protein

Marke:  R&D Systems™ 5836-FS-025/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13415564

  • 315.49 € / 25 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to neutralize Activin-mediated erythroid differentiation of K562 human chronic myelogenous leukemia cells. The ED 50 for this effect is 0.05-0.25 µg/mL in the presence of 7.5 ng/mL of recombinant human Activin A.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
31.7 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
10468
Follistatin 288
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos