missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human gp130 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

N.V.

Spezifikation

Konjugat Unconjugated
Gen-ID (Entrez) 3572
Molekulargewicht 68 kDa
Name gp130
Rekombinant Yes
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Recombinant Human gp130 Protein is derived from Sf 21 (baculovirus). The Recombinant Human gp130 Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Unconjugated
68 kDa
Yes
3572
gp130
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human gp130 protein Leu24-Glu619 (Glu619Asp)
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen