missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human gp130 Recombinant Protein

Marke:  R&D Systems™ 228-GP-050/CF

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13410954

  • 1374.65 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to inhibit IL-6-dependent proliferation of the 7TD1 mouse hybridoma cells in the presence of Recombinant Human IL-6 R alpha (Catalog # 227-SR ). The ED 50 for this effect is 0.5-2 µg/mL in the presence of 4 µg/mL recombinant human (rh) IL-6 R alpha and 25 pg/mL rhIL-6.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS and Trehalose.
68 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3572
gp130
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Yes
Videos