missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GST UbcH2/UBE2H Recombinant Protein

Marke:  R&D Systems™ E2-605-100

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414865

  • 322.09 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Contains an N-terminal GST (glutathione S-transferase) tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 7328
Regulatory Status: RUO

Spezifikation

Spezifikation

No Formulation Info in Bulk Inventory
GST UbcH2/UBE2H
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Yes
7328
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Unlabeled
Videos