missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human GST UbcH5b/UBE2D2 Recombinant Protein

Marke:  R&D Systems™ E2-620-100

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414885

  • 322.09 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Contains an N-terminal GST (glutathione S-transferase) tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7322
Regulatory Status: RUO

Spezifikation

Spezifikation

Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
GST UbcH5b/UBE2D2
100μg
Unlabeled
7322
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Yes
Videos