missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human HGF Recombinant Protein

Marke:  R&D Systems™ 294-HG-005

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412114

  • 487.52 € / 5 Mikrogramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Measured by its ability to induce IL-11 secretion by Saos-2 human osteosarcoma cells. Hjertner, O. et al. (1999) Blood 94:3883. The ED 50 for this effect is 0.1-0.5 ng/mL. The specific activity of Recombinant Human HGF is approximately 1.5 x 10 3 IU/µg, which is calibrated against Recombinant Human HGF WHO International Standard (NIBSC code: 96/564).
Spezifikation

Spezifikation

Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein.
Predicted MW: 53.7kDa ( alpha chain), 26kDa ( beta chain) Observed MW: 60-70kDa, non-reducing conditions 60kDa, and 33-34kDa, reducing conditions
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Disulfide-linked heterodimer
Yes
3082
HGF
5μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unlabeled
Reconstitute at 100μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Videos