missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human His6 HR6B/UBE2B Recombinant Protein

Marke:  R&D Systems™ E2-613-100

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13414875

  • 322.09 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: E. coli-derived, Contains an N-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation: No Formulation Info in Bulk Inventory
Entrez Gene ID: 7320
Regulatory Status: RUO

Spezifikation

Spezifikation

No Formulation Info in Bulk Inventory
His6 HR6B/UBE2B
100μg
Unlabeled
7320
95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Human
Yes
Videos