missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human ICAM-1/CD54 Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
N.V.
Spezifikation
Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS. |
---|---|
Gen-ID (Entrez) | 3383 |
Molekulargewicht | 76 kDa (monomer) |
Name | ICAM-1/CD54 Fc Chimera |
Quelle | Mouse myeloma cell line,NS0-derived human ICAM-1/CD54 protein, (N-terminus) Human ICAM-1 Gln28-Glu480 Accession &Num; CAA30051 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Beschreibung
The Recombinant Human ICAM-1/CD54 Fc Chimera Protein is derived from NS0. The Recombinant Human ICAM-1/CD54 Fc Chimera Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
76 kDa (monomer) | |
Mouse myeloma cell line,NS0-derived human ICAM-1/CD54 protein, (N-terminus) Human ICAM-1 Gln28-Glu480 Accession &Num; CAA30051 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
Disulfide-linked homodimer | |
Recombinant |
3383 | |
ICAM-1/CD54 Fc Chimera | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 12 weeks, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |