missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ICAM-1/CD54 Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 720-IC-050

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416104

  • 360.37 € / 50 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Gln28-Glu480
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 76 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3383
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
76 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse myeloma cell line, NS0-derived Human ICAM-1 Gln28-Glu480 Accession # CAA30051 IEGRMD Human IgG1 (Pro100-Lys330)
Unlabeled
3383
ICAM-1/CD54 Fc Chimera
50μg
Disulfide-linked homodimer
Yes
Videos