missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human ICAM-3/CD50 Fc Chimera Recombinant Protein

Marke:  R&D Systems™ 715-IC-200

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13416084

  • 544.72 € / 200 Mikrogramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Source: Mouse myeloma cell line, NS0-derived, Gln30-His485
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 76.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 3385
Regulatory Status: RUO

Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in PBS.
76.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Unlabeled
3385
ICAM-3/CD50 Fc Chimera
200μg
Disulfide-linked homodimer
Yes
Videos